5-Pyridin-2-yl-1,3,4-oxadiazol-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 5-(Pyridin-2-yl)-1,3,4-oxadiazol-2-amine

A480222-100MG A480222-1G A480222-250MG
76774-352EA 47.75 USD
76774-352 76774-354 76774-356
5-Pyridin-2-yl-1,3,4-oxadiazol-2-amine 97%
5-Pyridin-2-yl-1,3,4-oxadiazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₆N₄O
MW: 162.15 g/mol
MDL Number: MFCD01710636
CAS Number: 5711-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR