Ethyl-2-methyl-4,4,4-trifluoroacetoacetate 96% (by GC)

Supplier: Ambeed
Danger

Synonyms: Ethyl 4,4,4-Trifluoro-2-methyl-3-oxobutyrate

A478573-1G A478573-25G A478573-5G
76774-336EA 14.86 USD
76774-336 76774-338 76774-340
Ethyl-2-methyl-4,4,4-trifluoroacetoacetate 96% (by GC)
Ethyl-2-methyl-4,4,4-trifluoroacetoacetate

Precaución: for Research Use Only

Formula: C₇H₉F₃O₃
MW: 198.14 g/mol
MDL Number: MFCD00190635
CAS Number: 344-00-3
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR