tert-Butyl-5-bromo-3-formyl-1H-indole-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 5-bromo-3-formyl-1H-indole-1-carboxylate, 5-Bromo-1H-indole-3-carboxaldehyde, N-BOC protected

A477349-1G A477349-5G
76774-328EA 19.1 USD
76774-328 76774-330
tert-Butyl-5-bromo-3-formyl-1H-indole-1-carboxylate 95%
tert-Butyl-5-bromo-3-formyl-1H-indole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₄BrNO₃
MW: 324.1700 g/mol
MDL Number: MFCD05864691
CAS Number: 325800-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR