3-Amino-N,N-diethylbenzenesulfonamide 95%

Supplier: Ambeed
Warning

A474925-1G A474925-5G
76774-320EA 74 USD
76774-320 76774-322
3-Amino-N,N-diethylbenzenesulfonamide 95%
3-Amino-N,N-diethylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂S
MW: 228.3100 g/mol
MDL Number: MFCD02720457
CAS Number: 10372-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR