5-(2-Thioxo-2,3-dihydrothiazol-4-yl)thiophene-2-carboxamide ≥95%

Supplier: Ambeed
Warning

A474528-25G A474528-5G
76774-316EA 195 USD
76774-316 76774-318
5-(2-Thioxo-2,3-dihydrothiazol-4-yl)thiophene-2-carboxamide ≥95%
5-(2-Thioxo-2,3-dihydrothiazol-4-yl)thiophene-2-carboxamide

Precaución: for Research Use Only

Formula: C₈H₆N₂OS₃
MDL Number: MFCD11040250
CAS Number: 52560-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR