2,3-Difluoro-6-(trifluoromethyl)benzonitrile 95%

Supplier: Ambeed
Warning

A474247-1G A474247-5G
76774-312EA 75.33 USD
76774-312 76774-314
2,3-Difluoro-6-(trifluoromethyl)benzonitrile 95%
2,3-Difluoro-6-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₂F₅N
MW: 207.1 g/mol
MDL Number: MFCD06658270
CAS Number: 186517-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR