6-Ethyl-4-methoxy-2-pyranone 97%

Supplier: Ambeed
Warning

A473908-1G A473908-250MG A473908-5G
76774-302EA 91.24 USD
76774-302 76774-304 76774-306
6-Ethyl-4-methoxy-2-pyranone 97%
6-Ethyl-4-methoxy-2-pyranone

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MW: 154.163 g/mol
MDL Number: MFCD09991688
CAS Number: 106950-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR