4-Fluoro-2-methylbenzenesulfonamide 95%

Supplier: Ambeed
Warning

A472561-1G A472561-5G
76774-294EA 28.65 USD
76774-294 76774-296
4-Fluoro-2-methylbenzenesulfonamide 95%
4-Fluoro-2-methylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₈FNO₂S
MW: 189.2073 g/mol
MDL Number: MFCD02709831
CAS Number: 489-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR