Quinoline ethiodide 95%

Supplier: Ambeed
Danger

A470797-25G A470797-5G
76774-278EA 35 USD
76774-278 76774-280
Quinoline ethiodide 95%
Quinoline ethiodide

Precaución: for Research Use Only

Formula: C₁₁H₁₂IN
MDL Number: MFCD00041996
CAS Number: 634-35-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR