2-Fluoro-6-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A469916-10G A469916-25G A469916-5G A469916-1G
76774-268EA 16 USD
76774-268 76774-270 76774-272 77878-236
2-Fluoro-6-nitrobenzoic acid 97%
2-Fluoro-6-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FNO₄
MDL Number: MFCD01862079
CAS Number: 385-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR