[6-(Trifluoromethyl)-3-pyridinyl]methanol 98%

Supplier: Ambeed
Danger

Synonyms: 5-(Hydroxymethyl)-2-(trifluoromethyl)pyridine, [6-(Trifluoromethyl)pyridin-3-yl]methanol

A466756-1G A466756-250MG A466756-5G
76774-254EA 45.62 USD
76774-254 76774-256 76774-258
[6-(Trifluoromethyl)-3-pyridinyl]methanol 98%
[6-(Trifluoromethyl)-3-pyridinyl]methanol

Precaución: for Research Use Only

Formula: C₇H₆F₃NO
MW: 177.13 g/mol
MDL Number: MFCD01862648
CAS Number: 386704-04-7
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR