5-Nitro-2-thiophenecarbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 5-Nitrothiophene-2-carbonitrile

A463709-10G A463709-1G A463709-25G A463709-5G
76774-240EA 36.08 USD
76774-240 76774-242 76774-244 76774-246
5-Nitro-2-thiophenecarbonitrile 98%
5-Nitro-2-thiophenecarbonitrile

Precaución: for Research Use Only

Formula: C₅H₂N₂O₂S
MW: 154.15 g/mol
MDL Number: MFCD00052401
CAS Number: 16689-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR