Pyridine-4-sulfonic acid 96%

Supplier: Ambeed
Warning

Synonyms: 4-Pyridinesulphonic acid, 4-Sulphopyridine

A462542-100MG A462542-1G A462542-250MG A462542-5G
76774-232EA 14 USD
76774-232 76774-234 76774-236 77626-922
Pyridine-4-sulfonic acid 96%
Pyridine-4-sulfonic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₃S
MDL Number: MFCD00234082
CAS Number: 5402-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR