6-Fluoroindol-3-ylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(6-Fluoro-1H-indol-3-yl)acetic acid, 6-Fluoroindole-3-acetic acid

A462168-1G A462168-250MG A462168-5G
76774-228EA 60.48 USD
76774-228 76774-230 77626-918
6-Fluoroindol-3-ylacetic acid 98%
6-Fluoroindol-3-ylacetic acid

Precaución: for Research Use Only

Formula: C₁₀H₈FNO₂
MW: 193.18 g/mol
MDL Number: MFCD01074503
CAS Number: 443-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR