β-D-Allose 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol, β-D-Allopyranose, (2R,3R,4R,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol

A460994-1G A460994-250MG A460994-5G
76774-220EA 128 USD
76774-220 76774-222 76774-224
β-D-Allose 98%
β-D-Allose

Precaución: for Research Use Only

Formula: C₆H₁₂O₆
MDL Number: MFCD00063268
CAS Number: 7283-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR