cis-1,2-Dimethylcyclohexane 97%

Supplier: Ambeed
Danger

A460556-1G A460556-5G
76774-214EA 14 USD
76774-214 76774-216
cis-1,2-Dimethylcyclohexane 97%

Precaución: for Research Use Only

Formula: C₈H₁₆
MDL Number: MFCD00063621
CAS Number: 2207-01-4
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR