3-Benzylamino-1-propanol 96%

Supplier: Ambeed
Warning

Synonyms: 3-(Benzylamino)propan-1-ol, N-(3-Hydroxypropyl)benzylamine, 3-(Benzylamino)-1-propanol

A460287-100G A460287-10G A460287-1G A460287-25G A460287-5G
76774-204EA 192 USD
76774-204 76774-206 76774-208 76774-210 76774-212
3-Benzylamino-1-propanol 96%
3-Benzylamino-1-propanol

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO
MDL Number: MFCD00020622
CAS Number: 4720-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR