6-Azauridine 98%

Supplier: Ambeed
Danger

Synonyms: 2-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,2,4-triazine-3,5(2H,4H)-dione, 6-Azuridine

A458903-1MG A458903-1G A458903-250MG A458903-25G A458903-5G
76774-194EA 6 USD
76774-194 77601-984 77601-986 77601-988 77601-990
6-Azauridine 98%
6-Azauridine

Precaución: for Research Use Only

Formula: C₈H₁₁N₃O₆
MW: 245.19 g/mol
MDL Number: MFCD00006472
CAS Number: 54-25-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR