Boc-Leu-OSu 96%

Supplier: Ambeed
Warning

A458571-1G A458571-25G A458571-5G
76774-188EA 8.49 USD
76774-188 76774-190 76774-192
Boc-Leu-OSu 96%
Boc-Leu-OSu

Precaución: for Research Use Only

Formula: C₁₅H₂₄N₂O₆
MW: 328.365 g/mol
MDL Number: MFCD00037913
CAS Number: 3392-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR