6-Fluoro-1H-indazol-3(2H)-one 95%

Supplier: Ambeed
Warning

A458553-1G A458553-250MG
76774-184EA 192.03 USD
76774-184 76774-186
6-Fluoro-1H-indazol-3(2H)-one 95%
6-Fluoro-1H-indazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₇H₅FN₂O
MW: 152.1300 g/mol
MDL Number: MFCD16038756
CAS Number: 862274-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR