5-Bromo-2-iodopyridin-3-ol 97%

Supplier: Ambeed
Warning

A458523-100G A458523-100MG A458523-1G A458523-250MG A458523-25G A458523-5G
76774-172EA 1436.46 USD
76774-172 76774-174 76774-176 76774-178 76774-180 76774-182
5-Bromo-2-iodopyridin-3-ol 97%
5-Bromo-2-iodopyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃BrINO
MDL Number: MFCD09859112
CAS Number: 188057-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR