Cyclobutanecarbonitrile 95%

Supplier: Ambeed
Danger

A456926-10G A456926-1G A456926-25G A456926-5G
76774-164EA 68.96 USD
76774-164 76774-166 76774-168 76774-170
Cyclobutanecarbonitrile 95%
Cyclobutanecarbonitrile

Precaución: for Research Use Only

Formula: C₅H₇N
MW: 81.12 g/mol
MDL Number: MFCD00013735
CAS Number: 4426-11-3
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR