Ethyl-6-(hydroxymethyl)pyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-(hydroxymethyl)pyridine-2-carboxylate

A453169-100MG A453169-1G
76774-150EA 48.81 USD
76774-150 76774-152
Ethyl-6-(hydroxymethyl)pyridine-2-carboxylate 98%
Ethyl-6-(hydroxymethyl)pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD09750009
CAS Number: 41337-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR