4-Acetamido-2-aminobenzenesulfonic acid hydrate 95+%

Supplier: Ambeed
Warning

Synonyms: 3'-Amino-4'-sulphoacetanilide, 4-Acetamido-2-aminobenzenesulphonic acid

A451369-1G A451369-5G
76774-128EA 11.67 USD
76774-128 76774-130
4-Acetamido-2-aminobenzenesulfonic acid hydrate 95+%
4-Acetamido-2-aminobenzenesulfonic acid hydrate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₄S
MW: 230.24 g/mol
MDL Number: MFCD00035752
CAS Number: 88-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR