6-Fluorothio-4-chromanone 97%

Supplier: Ambeed
Warning

Synonyms: 6-Fluorothiochroman-4-one

A450044-1G A450044-25G A450044-5G
76774-114EA 16.98 USD
76774-114 76774-116 76774-118
6-Fluorothio-4-chromanone 97%
6-Fluorothio-4-chromanone

Precaución: for Research Use Only

Formula: C₉H₇FOS
MW: 182.22 g/mol
MDL Number: MFCD00078488
CAS Number: 21243-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR