2-(4-Amino-2-nitroanilino)ethanol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-N-(2-hydroxyethyl)-2-nitroaniline, N1-(2-Hydroxyethyl)-2-nitro-1,4-phenylenediamine, 2-((4-Amino-2-nitrophenyl)amino)ethanol

A449180-100G A449180-25G A449180-500G
76774-094EA 43 USD
76774-094 76774-096 76774-098
2-(4-Amino-2-nitroanilino)ethanol 98%
2-(4-Amino-2-nitroanilino)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₁N₃O₃
MDL Number: MFCD00071801
CAS Number: 2871-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR