(3R)-(-)-1-Benzyl-3-aminopyrrolidine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-1-Benzylpyrrolidin-3-amine, (R)-(-)-1-Benzyl-3-aminopyrrolidine

A448813-10G A448813-1G A448813-250MG A448813-5G
76774-080EA 161.26 USD
76774-080 76774-082 76774-084 76774-086
(3R)-(-)-1-Benzyl-3-aminopyrrolidine 97%
(3R)-(-)-1-Benzyl-3-aminopyrrolidine

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
MDL Number: MFCD00082638
CAS Number: 114715-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR