6-(Chloromethyl)pyridine-2-carbonitrile 97%

Supplier: Ambeed
Danger

Synonyms: 6-(Chloromethyl)-2-cyanopyridine

A444463-1G A444463-250MG A444463-5G
76774-070EA 29.71 USD
76774-070 76774-072 76774-074
6-(Chloromethyl)pyridine-2-carbonitrile 97%
6-(Chloromethyl)pyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD09991809
CAS Number: 135450-23-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR