Hexaminolevulinate hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Hexaminolevulinate HCl
A444075-100MG
A444075-1MG
A444075-25MG
A444075-250MG
A444075-1G
A444075-5G
76774-064EA
17
USD
76774-064
76774-066
76774-068
77626-890
77878-207
77878-208
Hexaminolevulinate hydrochloride 98%
Hexaminolevulinate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₂₂ClNO₃ MW: 251.75 g/mol |
MDL Number:
MFCD03695491 CAS Number: 140898-91-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...