3-Chloro-4-nitrotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-methyl-1-nitrobenzene

A444068-100G A444068-1G
76774-060EA 60.48 USD
76774-060 76774-062
3-Chloro-4-nitrotoluene 98%
3-Chloro-4-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD02683043
CAS Number: 38939-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR