Ethyl-2-thiopheneacetate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(thiophen-2-yl)acetate

A443158-100G A443158-1G A443158-25G A443158-5G
76774-052EA 78 USD
76774-052 76774-054 76774-056 76774-058
Ethyl-2-thiopheneacetate 95%
Ethyl-2-thiopheneacetate

Precaución: for Research Use Only

Formula: C₈H₁₀O₂S
MDL Number: MFCD00005457
CAS Number: 57382-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR