3-(Boc-amino)-4-methylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Boc-amino)-4-methylbenzeneboronic acid, 3-BOC-amino-4-methylphenylboronic acid, (3-((tert-Butoxycarbonyl)amino)-4-methylphenyl)boronic acid

A440078-1G A440078-250MG A440078-5G
76774-030EA 135.8 USD
76774-030 76774-032 76774-034
3-(Boc-amino)-4-methylphenylboronic acid 98%
3-(Boc-amino)-4-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₄
MW: 251.09 g/mol
MDL Number: MFCD04115654
CAS Number: 850568-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR