(±)-Ethyl-2-bromovalerate 97%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2-bromovalerate

A438796-25G A438796-5G
76774-026EA 18 USD
76774-026 76774-028
(±)-Ethyl-2-bromovalerate 97%
(±)-Ethyl-2-bromovalerate

Precaución: for Research Use Only

Formula: C₇H₁₃BrO₂
MDL Number: MFCD00000159
CAS Number: 615-83-8
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR