3-(Trifluoromethyl)phenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3-(Trifluoromethyl)phenyl)boronic acid, (3-(Trifluoromethyl)phenyl)metaboric acid, 3-(Trifluoromethyl)benzeneboronic acid, Dihydroxy(3-(trifluoromethyl)phenyl)borane
A437948-100G
A437948-10G
A437948-1G
A437948-25G
A437948-5G
A437948-500G
76774-016EA
95
USD
76774-016
76774-018
76774-020
76774-022
76774-024
77626-872
3-(Trifluoromethyl)phenylboronic acid 98%
3-(Trifluoromethyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₆BF₃O₂ |
MDL Number:
MFCD00151854 CAS Number: 1423-26-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...