5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) 97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dioxopyrrolidin-1-yl 3',6'-dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylate, 5-FAM-N-succinimidyl ester, 5-Carboxyfluorescein NHS ester, 5-FAM SE

A437441-1MG A437441-100MG A437441-250MG A437441-1G
76774-012EA 10.61 USD
76774-012 77551-312 77572-354 77601-890
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) 97%
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE)

Precaución: for Research Use Only

Formula: C₂₅H₁₅NO₉
MW: 473.39 g/mol
MDL Number: MFCD00077322
CAS Number: 92557-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR