5,6-Dimethoxybenzo[d]thiazol-2-amine 98%

Supplier: Ambeed
Warning

A435808-10G A435808-1G A435808-250MG A435808-5G
76773-988EA 919.81 USD
76773-988 76773-990 76773-992 76773-994
5,6-Dimethoxybenzo[d]thiazol-2-amine 98%
5,6-Dimethoxybenzo[d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₂S
MW: 210.25 g/mol
MDL Number: MFCD01663342
CAS Number: 6294-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR