Allyltriethoxysilane 95%

Supplier: Ambeed
Danger

A435376-1G A435376-25G A435376-5G
76773-978EA 10 USD
76773-978 76773-980 76773-982
Allyltriethoxysilane 95%
Allyltriethoxysilane

Precaución: for Research Use Only

Formula: C₉H₂₀O₃Si
MW: 204.34 g/mol
MDL Number: MFCD00009064
CAS Number: 2550-04-1
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR