(±)-1-Benzyl-3-hydroxypiperidine 98%

Supplier: Ambeed
Warning

Synonyms: N-Benzyl-3-hydroxypiperidine, 1-Benzylpiperidin-3-ol, 1-Benzyl-3-piperidinol, 1-Benzyl-3-hydroxypiperidine

A434216-1G A434216-500G A434216-5G
76773-966EA 9.55 USD
76773-966 76773-968 76773-970
(±)-1-Benzyl-3-hydroxypiperidine 98%
(±)-1-Benzyl-3-hydroxypiperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO
MW: 191.27 g/mol
MDL Number: MFCD00023734
CAS Number: 14813-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR