5-Bromo-1,2,4-trimethylbenzene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2,4,5-trimethylbenzene

A434130-100G A434130-10G A434130-1G A434130-5G A434130-25G
76773-958EA 118.83 USD
76773-958 76773-960 76773-962 76773-964 77626-864
5-Bromo-1,2,4-trimethylbenzene 97%
5-Bromo-1,2,4-trimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁Br
MW: 199.09 g/mol
MDL Number: MFCD00000072
CAS Number: 5469-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR