Ethyl-2,4-dihydroxy-6-methylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2,4-dihydroxy-6-methylbenzoate, 4-Ethoxycarbonyl-5-methylresorcinol, 2-Ethoxycarbonyl-3,5-dihydroxytoluene, 2-Carbethoxy-3,5-dihydroxytoluene, 4-Carbethoxy-5-methylresorcinol, 2,4-Dihydroxy-6-methylbenzoic Acid Ethyl Ester

A432422-1G A432422-1MG A432422-250MG A432422-50MG A432422-5G A432422-25G
76773-930EA 50.93 USD
76773-930 76773-932 76773-934 76773-936 76773-938 77626-862
Ethyl-2,4-dihydroxy-6-methylbenzoate 98%
Ethyl-2,4-dihydroxy-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MW: 196.20 g/mol
MDL Number: MFCD00010821
CAS Number: 2524-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR