2,4-Dinitro-5-fluorotoluene 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-Fluoro-5-methyl-2,4-dinitrobenzene
A432015-1G
A432015-250MG
76773-926EA
20.16
USD
76773-926
76773-928
2,4-Dinitro-5-fluorotoluene 95%
2,4-Dinitro-5-fluorotoluene
Precaución: for Research Use Only
Formula:
C₇H₅FN₂O₄ MW: 200.1240 g/mol |
MDL Number:
MFCD00205111 CAS Number: 349-01-9 UN: 1325 ADR: 4.1,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...