4-Bromo-N,N-diethylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

A430935-1G A430935-250MG A430935-5G
76773-920EA 28 USD
76773-920 76773-922 76773-924
4-Bromo-N,N-diethylbenzenesulfonamide 98%
4-Bromo-N,N-diethylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrNO₂S
MDL Number: MFCD00587586
CAS Number: 90944-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR