6-Bromo-1H-imidazo[4,5-b]pyridin-2(3H)-one 95%

Supplier: Ambeed
Warning

A430414-100MG A430414-10G A430414-1G A430414-250MG A430414-5G
76773-910EA 8 USD
76773-910 76773-912 76773-914 76773-916 76773-918
6-Bromo-1H-imidazo[4,5-b]pyridin-2(3H)-one 95%
6-Bromo-1H-imidazo[4,5-b]pyridin-2(3H)-one

Precaución: for Research Use Only

Formula: C₆H₄BrN₃O
MDL Number: MFCD00763285
CAS Number: 148038-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR