5-Methyl-1,3-benzodioxole 95%

Supplier: Ambeed
Danger

A428303-1G A428303-5G A428303-250MG A428303-25G
76773-900EA 43 USD
76773-900 77598-482 77878-179 77878-180
5-Methyl-1,3-benzodioxole 95%
5-Methyl-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₈H₈O₂
MDL Number: MFCD00005833
CAS Number: 7145-99-5
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR