5-Bromo-7-chloro-1H-indazole 98%

Supplier: Ambeed
Warning

A425872-100MG A425872-250MG A425872-5G A425872-1G
76773-886EA 49.87 USD
76773-886 76773-888 76773-890 77598-478
5-Bromo-7-chloro-1H-indazole 98%
5-Bromo-7-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.4771 g/mol
MDL Number: MFCD11044588
CAS Number: 635712-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR