(Bis(trifluoroacetoxy)iodo)pentafluorobenzene 97%

Supplier: Ambeed
Danger

A422899-1G A422899-5G
76773-872EA 96 USD
76773-872 77878-174
(Bis(trifluoroacetoxy)iodo)pentafluorobenzene 97%
(Bis(trifluoroacetoxy)iodo)pentafluorobenzene

Precaución: for Research Use Only

Formula: C₁₀F₁₁IO₄
MDL Number: MFCD00191612
CAS Number: 14353-88-9
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR