2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoropyridine-2-boronic acid pinacol ester

A422377-1G A422377-250MG A422377-5G
76773-866EA 312.97 USD
76773-866 76773-868 76773-870
2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%
2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
MDL Number: MFCD08063029
CAS Number: 842136-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR