6-Methylnaphthalen-2-ol 97%

Supplier: Ambeed
Warning

A419663-100MG A419663-1G A419663-250MG
76773-860EA 41 USD
76773-860 76773-862 76773-864
6-Methylnaphthalen-2-ol 97%
6-Methylnaphthalen-2-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O
MDL Number: MFCD00046473
CAS Number: 17579-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR