5-Bromo-3-fluorosalicylaldehyde 95%

Supplier: Ambeed
Warning

A417511-250MG A417511-1G A417511-25G
76773-858EA 7 USD
76773-858 77594-292 77626-836
5-Bromo-3-fluorosalicylaldehyde 95%
5-Bromo-3-fluorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
MDL Number: MFCD03788530
CAS Number: 251300-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR