4-(Bromomethyl)-1-fluoro-2-nitrobenzene 98%

Supplier: Ambeed
Danger

A415873-10G A415873-1G A415873-25G A415873-5G
76773-844EA 31 USD
76773-844 76773-846 76773-848 76773-850
4-(Bromomethyl)-1-fluoro-2-nitrobenzene 98%
4-(Bromomethyl)-1-fluoro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD03412241
CAS Number: 15017-52-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR